Chloroform-[D₁] (99.8% D) for NMR spectroscopy

Supplier: VWR International
Danger

BDH0904-100ML BDH0904-10ML BDH0904-25ML
BDH0904-100MLEA 52.88 USD
BDH0904-100ML BDH0904-10ML BDH0904-25ML
Chloroform-[D₁] (99.8% D) for NMR spectroscopy
Chloroform-[D1]

Deuterium Labeled Compounds. These high-quality products are used as solvents in NMR spectroscopy, R&D, tracers, and building blocks in pharmaceutical ingredients, as well as for the synthesis of organic light-emitting diodes (OLED).


  • Highest quality deuterated stable isotopes for NMR. NMR solvents are batch tested for both chemical and isotopic purity by 1H-NMR, GC and Karl Fischer titration.

Formula: CDCl₃
Boiling Pt: 61.5 °C (1013 hPa)
Melting Pt: –64.7 … –64.2 °C (4 hPa)
Density: 1.45 g/cm³ (20 °C)
Flash Pt: > 60 °C (1019 hPa)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Appearance/color Colorless
Appearance/form Liquid
D content 1H-NMR min. 99.80 %(mol)
w(H2O) 1H-NMR max. 0.010 %(w)
1H-NMR Purity conform
H2O + D2O Content by KF Coulometer max 0.020 %(w)
GC Content min. 99.0 %(w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR