Magnesium (Mg2+) Single-Element Ion Cation Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR International
BDH0829-125ML
BDH0829-125MLEA
87.52
USD
BDH0829-125ML
Magnesium (Mg2+) Single-Element Ion Cation Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards
Cationic Standards
Magnesium Standards
Ion chromatography & ion selective electrode (ISE) standard.
- Single and multi-element standards for ion chromatography available
- Stable and traceable