Calcium (Ca2+) Single-Element Ion Cation Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Supplier: VWR International
BDH0827-125ML
BDH0827-125MLEA 87.52 USD
BDH0827-125ML
Calcium (Ca2+) Single-Element Ion Cation Standard, 1,000 µg/ml (1,000 ppm), VWR Chemicals BDH®
Standards Cationic Standards Calcium Standards
Ion chromatography & ion selective electrode (ISE) standard.
  • Single and multi-element standards for ion chromatography available
  • Stable and traceable