Dichloroacetic acid (DCA) 3% in anhydrous dichloromethane deblocking reagent, for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: DCA Deblock, DCA

BDH0609-2.5L BDH0609-200L BDH0609-4L BDH0609-56L
BDH0609-2.5LCS 412.94 USD
BDH0609-2.5L BDH0609-200L BDH0609-4L BDH0609-56L
Dichloroacetic acid (DCA) 3% in anhydrous dichloromethane deblocking reagent, for biosynthesis, VWR Chemicals BDH®
Dichloroacetic acid (DCA)

BDH Biosynthesis grade reagents are designed for biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.


  • Custom blends and bulk sizes available upon request.

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2922
ADR: 8 II
Merck Index: 13,03075

Order Now

Specification Test Results

Description (clear liquid free of suspended matter) Pass
Dichloroacetic Acid 2.95 - 3.05%
Color (at time of packaging) <10 APHA
Water Max 100 ppm
Dichloromethane (component purity) Min 99.9%
Dicloroacetic Acid (component purity) Min 98.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR