Dichloroacetic acid (DCA) 10% in anhydrous toluene deblocking reagent, for biosynthesis, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: DCA Deblock, DCA

BDH0605-2.5L BDH0605-200L BDH0605-4L BDH0605-56L
BDH0605-2.5LCS 1094.95 USD
BDH0605-2.5L BDH0605-200L BDH0605-4L BDH0605-56L
Dichloroacetic acid (DCA) 10% in anhydrous toluene deblocking reagent, for biosynthesis, VWR Chemicals BDH®
Dichloroacetic acid (DCA)

Deblock Solution (10% DCA in Toluene, v/v). BDH Biosynthesis Grade Reagents are designed for Biosynthesis research and production. Sizing available to fit your needs, from benchtop synthesizers to bulk production.


  • Custom Blends and Bulk Sizes available upon request.

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2924
ADR: 8 II
Merck Index: 13,03075

Order Now

Specification Test Results

Description (Clear liquid free of suspended matter) Passes test
Dichloroacetic acid 9.5 to 10.5 % Vol
Color (at time of packaging) Max 10 APHA
Water (by KF coulometry) Max 150 ppm
Dichloroacetic acid (component purity) Min 98.0 %
Toluene (component purity) Min 99.9 %
Titrable Acid or Base 89.5 to 90.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR