BD™ Quincke Spinal Needles, BD Medical
Supplier: BD
405162 405164 405174 405184 405171 405180 405182 400106 405073 405078 405081 405148 405149 405172 405071 405161 405181 408360 405074 405170
BD5162CS 648.2 USD
BD5162 BD405164 BD405174 BD405184 BD405211 BD5171 BD5180 BD5182 BD400106 BD405073 BD405078 BD405081 BD405148 BD405149 BD405172 BD5071 BD5161 BD405181 BD408360 BD5074 BD5170
BD™ Quincke Spinal Needles, BD Medical
Syringe Needles
Sterile, single-use needles have a unique key/slot arrangement of stylet and cannula hubs that facilitates proper needle bevel orientation
The translucent window hub allows for clear visualization of CSF, and fitted stylets reduce tissue coring