Corning® BioCoat® Poly-D-Lysine/Laminin 8-well Culture Slide, Corning
Supplier: Corning
354688
BD354688CS 476.41 USD
BD354688
Corning® BioCoat® Poly-D-Lysine/Laminin 8-well Culture Slide, Corning
Microplates Cell Culture Plates
Corning® BioCoat™ 8-well culture slide with a uniform application of PDL/Laminin. Glass slide with polystyrene vessel, lid, and safety removal tool. 4/pack, 12/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
  • Shelf life: >= 3 months from date of shipment