Corning® BioCoat™ FluoroBlok™ Fibronectin Cell Culture Inserts, Corning
Supplier: Corning
354597
BD354597CS 457.24 USD
BD354597
Corning® BioCoat™ FluoroBlok™ Fibronectin Cell Culture Inserts, Corning
Microplates Insert Plates and Inserts
Corning® BioCoat™ FluoroBlok™ fibronectin-coated cell culture inserts packaged ready to use in 24-well Falcon® companion plates.
  • Convenient, individual inserts packaged in a one-piece, automation friendly 24-multiwell format
  • Combined benefits of Falcon® inserts coated with HFN for enhanced cell attachment and growth
  • Individual inserts contain a 3.0 µm pore size clear PET membrane