Corning® BioCoat® Collagen I German Glass Coverslip, 22 mm, Bulk Packed, Corning
Supplier: Corning
354089
BD354089CS 199.65 USD
BD354089
Corning® BioCoat® Collagen I German Glass Coverslip, 22 mm, Bulk Packed, Corning
Cover Glasses
Corning® BioCoat™ 22 mm #1 German Glass coverslip with a uniform application of rat tail collagen. Bulk packed, 60/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
  • Shelf life: >= 3 months from date of shipment