Corning® BioCoat® Fibronectin German Glass Coverslip, 22 mm, Corning
Supplier: Corning
354088
BD354088CS 257.35 USD
BD354088
Corning® BioCoat® Fibronectin German Glass Coverslip, 22 mm, Corning
Cover Glasses
Corning® BioCoat™ 22×22 mm #1 German Glass coverslip with a uniform application of human fibronectin. Bulk packed, 60/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
  • Shelf life: >= 3 months from date of shipment