Corning® BioCoat® Poly-L-Lysine, German Glass Coverslip, 12 mm, Corning
Supplier: Corning
354085
BD354085CS 261.24 USD
BD354085
Corning® BioCoat® Poly-L-Lysine, German Glass Coverslip, 12 mm, Corning
Cover Glasses
Corning® BioCoat™ 12 mm #1 German Glass coverslip with a uniform application of Poly-L-Lysine. 40/pack, 80/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
  • Shelf life: >= 3 months from date of shipment