BD Taxo™ TB Niacin Test Control, BD Diagnostic Systems
Supplier: BD
231741
BD231741EA
199.2
USD
BD231741
BD Taxo™ TB Niacin Test Control, BD Diagnostic Systems
Controls and Calibration Verification Materials
BD Taxo™ TB Niacin Test Controls are impregnated with nicotinamide and give a positive reaction when tested with a TB Niacin Test Strip.