Vacutainer® Rapid Serum Tube with Hemogard™ Closure, BD Medical
Supplier: BD
368774
BD-368774CS 2512.41 USD
BD-368774
Vacutainer® Rapid Serum Tube with Hemogard™ Closure, BD Medical
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
BD Vacutainer® Rapid Serum Tubes (RST) contain thrombin-based clot activator and polymer gel for serum separation.
Samples processed in these tubes are used for serum determinations in chemistry. A five-minute clotting time makes this tube ideal for STAT testing in the emergency department as well as clinical laboratories striving to improve test turnaround and workflow efficiencies. This sterile, disposable plastic rapid serum tubes have paper label with orange hemogard closure to use for blood collection. Latex free.