Falcon® Feeder tray, 96-Well, Corning
Supplier: Corning
353924
BD-353924CS 155.87 USD
BD-353924
Falcon® Feeder tray, 96-Well, Corning
Trays/Dishes/Pots Laboratory Trays/Dishes
B DFalcon® HTS 96-multiwell feeder tray and lid, nontreated, hydrophobic polystyrene surface.
  • Sterile
  • Nonpyrogenic
For use with BD Falcon HTS FluoroBlok* 96-multiwell insert systems (BD351161, BD351162, BD351163, BD351164).