Fmoc-L-Tyr(Me)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine
4042449.0001
4042449.0005
B-4255.0001BAEA
230.72
USD
B-4255.0001BA
B-4255.0005BA
Fmoc-L-Tyr(Me)-OH
Fmoc-L-Tyr(Me)-OH
Building block for oxytocin and vasopressin analogs as carbetocin, and for structure-activity studies.
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine.Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Formula:
C₂₅H₂₃NO₅ MW: 417.47 g/mol |
MDL Number:
MFCD00153368 CAS Number: 77128-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...