Fmoc-Asp(Edans)-OH

Supplier: BACHEM AMERICAS INC

4049431.0500 4049431.1000
B-3785.0500BAEA 702.63 USD
B-3785.0500BA B-3785.1000BA
Fmoc-Asp(Edans)-OH
Fmoc-Asp(Edans)-OH

Sequence: Fmoc-Asp(EDANS)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Building block for the synthesis of internally quenched fluorogenic peptide substrates. The fluorescent group EDANS can be efficiently quenched by the nonfluorescent DABCYL chromophore.

Formula: C₃₁H₂₉N₃O₈S
Storage Temperature: Freezer
MDL Number: MFCD02094140
CAS Number: 182253-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR