N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH

4018282.0005 4018282.0025
B-2485.0005BAEA 114.5 USD
B-2485.0005BA B-2485.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid

Sequence: Fmoc-Asp-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


The oxazolidinone obtained from Fmoc-Asp-OH, paraformaldehyde and TosOH as catalyst has been converted into Dap derivatives, β-diazomethyl ketones, N-glycosylated Asn, and N'-glycosylated Dap Nβ-ureas. Heating in acetanhydride yields the cyclic anhydride which could be converted rather regioselectively into the β-anilide with o-anisidine and into N-glycosylated Asn with protected glycosylamines in DMSO.

Formula: C₁₉H₁₇NO₆
MW: 355.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237654
CAS Number: 119062-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR