Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine

4014948.0005 4014948.0025 4014948.0100
B-1785.0005BAEA 113.97 USD
B-1785.0005BA B-1785.0025BA B-1785.0100BA
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine

Sequence: Fmoc-Asn(Trt)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₃₈H₃₂N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00077049
CAS Number: 132388-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR