N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Supplier: BACHEM AMERICAS INC
4005510.0005
4005510.0025
B-1020.0005BAEA
158.5
USD
B-1020.0005BA
B-1020.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Sequence: Fmoc-D-Ala-OH · H₂O
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
Formula:
C₁₈H₁₇NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00062960 CAS Number: 79990-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...