N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate

Supplier: BACHEM AMERICAS INC

4005510.0005 4005510.0025
B-1020.0005BAEA 158.5 USD
B-1020.0005BA B-1020.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate

Sequence: Fmoc-D-Ala-OH · H₂O


Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).

Formula: C₁₈H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00062960
CAS Number: 79990-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR