Keto-Diastix® Reagent Strips for Urinalysis, Ascensia Diabetes Care
Supplier: ASCENSIA DIABETES CARE
2883
AM2883CS 679.55 USD
AM2883
Keto-Diastix® Reagent Strips for Urinalysis, Ascensia Diabetes Care
Clinical Diagnostic Supplies Clinical Reagents and Kits
Keto-Diastix® is a ready-to-use strip with two reagent areas specifically designed to provide a fast, convenient way to test urine for glucose and ketone.
  • Easy dip testing for urine glucose
  • Accurate timing is essential to provide reliable results
When the strip is dipped into a fresh urine sample one area of the strip will change color if it detects glucose. The other area of the strip will change color if it detects ketones. The colors will vary depending on the amount of each substance found in the urine.