Aluminium oxide nanoparticles 20% (w/w) in water 30 nm (TEM)

Supplier: ALADDIN SCIENTIFIC

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

A283857-100ML
ALA283857100MLEA 46.46 USD
ALA283857100ML
Aluminium oxide nanoparticles 20% (w/w) in water 30 nm (TEM)
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2977°C
Melting Pt: 2000°C
Density: 1.06 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf79c8baba0660bddb0ee84174dab6b396b87bd&vid=6cf79c8baba0660bddb0ee84174dab6b396b87bd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR