Aluminium chloride ≥99.99% (metals basis), PrimorTrace™, Electronic Grade

Supplier: ALADDIN SCIENTIFIC
Danger

A283582-25G
ALA283582-25GEA 379.91 USD
ALA283582-25G
Aluminium chloride ≥99.99% (metals basis), PrimorTrace™, Electronic Grade
Aluminium chloride

It is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

Formula: AlCl₃
Boiling Pt: 182.7°C
Melting Pt: 190°C
Storage Temperature: Ambient
MDL Number: MFCD00003422
CAS Number: 7446-70-0
EINECS: 231-208-1
UN: 1726
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR