AK-7 ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-(Azepan-1-ylsulphonyl)-N-(3-bromophenyl)benzamide, 3-(Azepan-1-ylsulfonyl)-N-(3-bromophenyl)benzamide

A274824-100MG A274824-25MG A274824-50MG A274824-5MG
ALA274824100MGEA 676.31 USD
ALA274824100MG ALA274824-25MG ALA274824-50MG ALA274824-5MG
AK-7 ≥98%
AK-7

AK-7 is a selective cell- and brain-permeable SIRT2 inhibitor, with an IC50 of 15.5 μM.

Formula: C₁₉H₂₁BrN₂O₃S
Storage Temperature: Freezer
MDL Number: MFCD03140195
CAS Number: 420831-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f2b3b4835eb441844d3aab02f2d32fdfc300b40&vid=2f2b3b4835eb441844d3aab02f2d32fdfc300b40&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR