AK-7 ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-(Azepan-1-ylsulphonyl)-N-(3-bromophenyl)benzamide, 3-(Azepan-1-ylsulfonyl)-N-(3-bromophenyl)benzamide
A274824-100MG
A274824-25MG
A274824-50MG
A274824-5MG
ALA274824100MGEA
676.31
USD
ALA274824100MG
ALA274824-25MG
ALA274824-50MG
ALA274824-5MG
AK-7 ≥98%
AK-7
AK-7 is a selective cell- and brain-permeable SIRT2 inhibitor, with an IC50 of 15.5 μM.
Formula:
C₁₉H₂₁BrN₂O₃S Storage Temperature: Freezer |
MDL Number:
MFCD03140195 CAS Number: 420831-40-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...