Agar, Bacteriological Grade

Supplier: ALADDIN SCIENTIFIC

Synonyms: Agar Agar

A274231-500G
ALA274231-500GEA 326.71 USD
ALA274231-500G
Agar, Bacteriological Grade
Agar

solidifying agent used in bacteriology. Agar consists of unbranched polysaccharides of galactose subunits derived from algae or seaweed.
Agar for the growth of bacteria and fungi
Typically used at concentrations of 1-2% in solid media preparation

Formula: C₁₂nH₁₈nOn₉n
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3841e3b8dfc2508c51689ae8e54203d8ede79dc1&vid=3841e3b8dfc2508c51689ae8e54203d8ede79dc1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR