Diammonium thiosulfate ≥96%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Ammonium hyposulphite, Ammonium hyposulfite, Ammonium thiosulphate, Ammonium thiosulfate

A264876-25G
ALA264876-25GEA 12.26 USD
ALA264876-25G
Diammonium thiosulfate ≥96%
di-Ammonium thiosulphate

Ammonium thiosulfate is a non-toxic inorganic compound that is majorly used as a fertilizer and can be used as an alternative to cyanide based leaching. It has a hygroscopic nature and is soluble in water. It can be produced by the reaction of ammonia, water, and sulfur dioxide. It can be used as a photographic fixing salt.Ammonium thiosulfide can be used as a source of sulfur for the formation of cadmium sulfide thin films for electronic applications.It can also be used in the treatment of copper-gold ores for the recovery of gold.

Formula: H₈N₂O₃S₂
MW: 148.21 g/mol
Density: 1.679 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00015965
CAS Number: 7783-18-8
EINECS: 231-982-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eda8d6d1f003548983888755991385d0293d0c1a&vid=eda8d6d1f003548983888755991385d0293d0c1a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR