3-Aminobutan-1-ol ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Amino-butan-1-ol

A183438-25G A183438-5G A183438-100G A183438-1G
ALA183438-25GEA 67.36 USD
ALA183438-25G ALA183438-5G ALA183438-100G ALA183438-1G
3-Aminobutan-1-ol ≥98%
3-Aminobutan-1-ol
Formula: C₄H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD03412694
CAS Number: 2867-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d04739695656ab6d1a77fa32ed2920c53dcf815f&vid=d04739695656ab6d1a77fa32ed2920c53dcf815f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR