3-(1-(4-Aminobutyl)-1H-1,2,3-triazol-4-yl)aniline trihydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC

A175084-250MG
ALA175084250MGEA 13.21 USD
ALA175084250MG
3-(1-(4-Aminobutyl)-1H-1,2,3-triazol-4-yl)aniline trihydrochloride ≥97%
3-(1-(4-Aminobutyl)-1H-1,2,3-triazol-4-yl)aniline trihydrochloride
Formula: C₁₂H₂₀Cl₃N₅
Storage Temperature: Ambient
MDL Number: MFCD28502706
CAS Number: 1788044-10-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b09a64ce37dd5f235e4c885330773254b56904ae&vid=b09a64ce37dd5f235e4c885330773254b56904ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR