2-Amino-5-hydroxy-1,3-benzothiazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A172351-1G
ALA172351-1GEA 464.46 USD
ALA172351-1G
2-Amino-5-hydroxy-1,3-benzothiazole ≥97%
2-Amino-5-hydroxy-1,3-benzothiazole
Formula: C₇H₆N₂OS
Storage Temperature: Refrigerator
MDL Number: MFCD00985125
CAS Number: 118526-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=addd953a652fbcf872fa43192d183d77c7652139&vid=addd953a652fbcf872fa43192d183d77c7652139&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR