5-Amino-3-(4-chlorophenyl)isoxazole ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 3-(4-Chlorophenyl)isoxazol-5-amine

A169693-1G
ALA169693-1GEA 161.41 USD
ALA169693-1G
5-Amino-3-(4-chlorophenyl)isoxazole ≥97%
5-Amino-3-(4-chlorophenyl)isoxazole
Formula: C₉H₇ClN₂O
MW: 194.6177 g/mol
Melting Pt: 163-167°C
Storage Temperature: Ambient
MDL Number: MFCD01569450
CAS Number: 33866-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4f655b3b3a490e555cb1d57b39e0568614ba3ee&vid=f4f655b3b3a490e555cb1d57b39e0568614ba3ee&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR