5-Amino-2,4-difluorophenol ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-amino-2,4-difluorophenol
A166012-100MG
A166012-1G
A166012-250MG
A166012-25G
A166012-50MG
A166012-5G
ALA166012100MGEA
79.71
USD
ALA166012100MG
ALA166012-1G
ALA166012250MG
ALA166012-25G
ALA166012-50MG
ALA166012-5G
5-Amino-2,4-difluorophenol ≥98%
5-Amino-2,4-difluorophenol
Formula:
C₆H₅F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD04973718 CAS Number: 113512-71-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...