Azido PEG Biotin

Supplier: ALADDIN SCIENTIFIC
Warning

A163258-200MG
ALA163258200MGEA 326.71 USD
ALA163258200MG
Azido PEG Biotin
Azido PEG Biotin

Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.

Formula: NH₂(CH₂CH₂O)nH
Storage Temperature: Ambient
CAS Number: 32130-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR