Amino PEG ≥95%
Supplier: ALADDIN SCIENTIFIC
A163244-200MG
ALA163244200MGEA 125.31 USD
ALA163244200MG
Amino PEG ≥95%
Amino PEG
Amino functionalized polyethylene glycol (NH2-PEG-NH2) is a homobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via carboxylic group (-COOH) or other amine reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.