1-Amino-2-bromonaphthalene ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151779-250MG
ALA151779250MGEA 28.41 USD
ALA151779250MG
1-Amino-2-bromonaphthalene ≥98%
1-Amino-2-bromonaphthalene
Formula: C₁₀H₈BrN
Melting Pt: 66.0-70.0 °C
Storage Temperature: Ambient
MDL Number: MFCD11870111
CAS Number: 771-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc53d926c2196e06f163c00d5eb440895a4c5042&vid=fc53d926c2196e06f163c00d5eb440895a4c5042&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR