5-Amino-3-methylisoxazole ≥97% (by titration)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine
A151656-5G
A151656-1G
ALA151656-5GEA
93.01
USD
ALA151656-5G
ALA151656-1G
5-Amino-3-methylisoxazole ≥97% (by titration)
5-Amino-3-methylisoxazole
5-Amino-3-methylisoxazole reacts with α,β-unsaturated ketones to yield the corresponding isoxazolo[5,4-b]pyridines.
Formula:
C₄H₆N₂O MW: 98.1 g/mol Boiling Pt: 194 Melting Pt: 84 |
MDL Number:
MFCD00003151 CAS Number: 14678-02-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...