4-Anilino-1,1':4',1''-terphenyl ≥98%

Supplier: ALADDIN SCIENTIFIC

A151640-1G
ALA151640-1GEA 217.46 USD
ALA151640-1G
4-Anilino-1,1':4',1''-terphenyl ≥98%
4-Anilino-1,1':4',1''-terphenyl
Formula: C₂₄H₁₉N
Melting Pt: 193-197°C
MDL Number: MFCD28963769
CAS Number: 897671-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=372bf4370ec242bfdf59e7b590dbffa383d44457&vid=372bf4370ec242bfdf59e7b590dbffa383d44457&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR