(±)-α-Terpineol ≥95% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A151628-100ML
ALA151628100MLEA 36.96 USD
ALA151628100ML
(±)-α-Terpineol ≥95% (by GC)
(±)-α-Terpineol

α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity in Melaleuca alternifolia (tea tree) essential oil.[1] The reaction rate constant of α-terpineol with the OH radical and ozone was found to be (1.9±0.5)×10-10cm3 molecule-1s-1 and (3.0±0.2)×10-16cm3 molecule-1s-1, respectively.Application:α-Terpineol has been used as a reference standard for the identification of α-terpineol in tequila by solid phase microextraction (SPME) and capillary gas chromatography−mass spectrometry (GC-MS), and in virgin olive oil samples by headspace SPME followed by GC-MS equipped with flame ionization detector (FID).It may be used as a reference standard for the analysis of α-terpineol as trace volatile compound in wine by solid-phase extraction (SPE) followed by GC-MS and in essential oils and related compounds from aromatic plants by SPME and GC-MS.α-Terpineol may be used in the synthesis of α-terpinyl esters of acetic acid and acetic anhydride via lipase-mediated esterification.

Formula: C₁₀H₁₈O
Boiling Pt: 210-219℃
Melting Pt: 31-32°C
Density: 0.93 g/cm³
Flash Pt: 90°C
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS Number: 98-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6b8e2f6a41de9c8d11393275ce578119b4c0ce5&vid=c6b8e2f6a41de9c8d11393275ce578119b4c0ce5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR