4-Amino-2,2,6,6-tetramethylpiperidine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151605-100ML
A151605-25ML
A151605-5ML
ALA151605100MLEA
175.66
USD
ALA151605100ML
ALA151605-25ML
ALA151605-5ML
4-Amino-2,2,6,6-tetramethylpiperidine ≥98% (by GC)
4-Amino-2,2,6,6-tetramethylpiperidine
Formula:
C₉H₂₀N₂ Boiling Pt: 188-189°C Melting Pt: 16-18 °C Density: 0.912 g/cm³ Flash Pt: 75 °C |
MDL Number:
MFCD00005984 CAS Number: 36768-62-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...