4-Amino-2,2,6,6-tetramethylpiperidine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151605-100ML A151605-25ML A151605-5ML
ALA151605100MLEA 175.66 USD
ALA151605100ML ALA151605-25ML ALA151605-5ML
4-Amino-2,2,6,6-tetramethylpiperidine ≥98% (by GC)
4-Amino-2,2,6,6-tetramethylpiperidine
Formula: C₉H₂₀N₂
Boiling Pt: 188-189°C
Melting Pt: 16-18 °C
Density: 0.912 g/cm³
Flash Pt: 75 °C
MDL Number: MFCD00005984
CAS Number: 36768-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa6f1ea6b14592aef9586e5acecb56f288b6ef17&vid=aa6f1ea6b14592aef9586e5acecb56f288b6ef17&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR