4-Amino-2,3,5,6-tetrafluorobenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-2,3,5,6-tetrafluorobenzoic Acid

A151459-1G
ALA151459-1GEA 68.31 USD
ALA151459-1G
4-Amino-2,3,5,6-tetrafluorobenzoic acid ≥97%
4-Amino-2,3,5,6-tetrafluorobenzoic acid

Application:It is used in chemical research and as pharmaceutical intermediate.

Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Melting Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00007647
CAS Number: 944-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62befc3c5ca44f4f05d2fe086a0eab6a6d0a6add&vid=62befc3c5ca44f4f05d2fe086a0eab6a6d0a6add&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR