25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: O-(2-Azidoethyl)-O'-methylheptaethylene Glycol, Methyl-PEG8-Azide, mPEG8-N3, mPEG8-Azide

A151379-25MG
ALA151379-25MGEA 19.86 USD
ALA151379-25MG
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90% (by HPLC)
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane
Formula: C₁₇H₃₅N₃O₈
MW: 409.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13184962
CAS Number: 869718-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b41b0e48cac2b2e7469105995f658a8e6cc26d5&vid=1b41b0e48cac2b2e7469105995f658a8e6cc26d5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR