2-Amino-6-methoxybenzothiazole 98

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Methoxybenzothiazol-2-ylamine

A151205-100G
ALA151205-100GEA 36.96 USD
ALA151205-100G
2-Amino-6-methoxybenzothiazole 98
2-Amino-6-methoxybenzothiazole

2-Amino-6-methoxybenzothiazole undergoes Sandmeyer reaction on heating with isoamyl nitrite and CuBr2 to yield 2-bromo-6-methoxybenzothiazole.2-Amino-6-methoxybenzothiazole was used as building block in the syntheses of novel series of Schiff bases and 4-thiazolidinones. It was used in the synthesis of 2-cyano-6-methoxybenzothiazole, key intermediate for the synthesis of firefly luciferin.

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Boiling Pt: 348
Melting Pt: 160
MDL Number: MFCD00005787
CAS Number: 1747-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83d460c5424e0ac53aa164e35b74282eb9350c32&vid=83d460c5424e0ac53aa164e35b74282eb9350c32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR