8-Acetyl-7-hydroxy-4-methylcoumarin ≥98% (by GC)

Supplier: Aladdin Scientific
Warning

A151116-1G
ALA151116-1GEA 41.71 USD
ALA151116-1G
8-Acetyl-7-hydroxy-4-methylcoumarin ≥98% (by GC)
8-Acetyl-7-hydroxy-4-methylcoumarin
Formula: C₁₂H₁₀O₄
Melting Pt: 170 °C
CAS Number: 2555-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b34705c611fbfc573130ae4ea1c342b38a0eedcf&vid=b34705c611fbfc573130ae4ea1c342b38a0eedcf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR