2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl Azide, 2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate

A151074-1G A151074-250MG A151074-5G
ALA151074-1GEA 112.96 USD
ALA151074-1G ALA151074250MG ALA151074-5G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98% (by HPLC)
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl azide
Formula: C₁₄H₂₀N₄O₈
Melting Pt: 167°C
Storage Temperature: Freezer
MDL Number: MFCD00216968
CAS Number: 6205-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56c4457ee3f2379ebe29d46d22d8ed68e1d68800&vid=56c4457ee3f2379ebe29d46d22d8ed68e1d68800&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR