2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
Synonyms:
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl Azide, 2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate
A151074-1G
A151074-250MG
A151074-5G
ALA151074-1GEA
112.96
USD
ALA151074-1G
ALA151074250MG
ALA151074-5G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide ≥98% (by HPLC)
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl azide
Formula:
C₁₄H₂₀N₄O₈ Melting Pt: 167°C Storage Temperature: Freezer |
MDL Number:
MFCD00216968 CAS Number: 6205-69-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...