Acetyl chloride

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Acetyl chloride 1M in Dichloromethane

A140638-100ML
ALA140638100MLEA 25.56 USD
ALA140638100ML
Acetyl chloride
Acetyl chloride
Formula: C₂H₃ClO
Boiling Pt: 52°C
Melting Pt: -112°C
Density: 1.294 g/cm³
Flash Pt: 4.4°C (cc)
Storage Temperature: Refrigerator
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 3286
ADR: 3(8),II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7de655fc009c140b23f655fc276ec1f87bbc7ef9&vid=7de655fc009c140b23f655fc276ec1f87bbc7ef9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR