Aluminium oxide ≥99.9% (metals basis)

Supplier: ALADDIN SCIENTIFIC

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

A140296-100G A140296-500G
ALA140296-100GEA 9.41 USD
ALA140296-100G ALA140296-500G
Aluminium oxide ≥99.9% (metals basis)
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2977°C
Melting Pt: 2000°C
Density: 1.06 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e65a4931af7c8240eedb4ca2fc5f0eee908e4304&vid=e65a4931af7c8240eedb4ca2fc5f0eee908e4304&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR