Alizarin complexone ≥95%
Supplier: Aladdin Scientific
A140206-1G
A140206-25G
A140206-5G
ALA140206-1GEA
66.41
USD
ALA140206-1G
ALA140206-25G
ALA140206-5G
Alizarin complexone ≥95%
Alizarin complexone
Formula:
C₁₉H₁₅NO₈ Melting Pt: 190°C Density: 1.4109 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00001202 CAS Number: 3952-78-1 EINECS: 223-544-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...