Aflatoxin B2 ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A140008-1MG
ALA140008-1MGEA 587.96 USD
ALA140008-1MG
Aflatoxin B2 ≥98% (by HPLC)
Aflatoxin B2

8,9-dihidro derivate of aflatoxin B1. Apotent hepatotoxic and hepatocarcinogenic mycotoxinapplication:Aflatoxin B2 may be used as a reference standard for the determination of aflatoxin B2 in animal feed by high performance liquid chromatography with fluorescence detection (HPLC-FLD), as well as quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction method combined with ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS), in eggs by QuEchERS extraction method followed by analysis using UHPLC with tandem mass spectrometry (MS/MS) operating on electrospray-ionization interface (ESI) mode, in Brazil nuts by immunoaffinity column (IAC) and clean-up HPLC-FLD and in plant-based milk and dairy products by dispersive liquid-liquid microextraction (DLLME), QuEchERS extraction method and HPLC-FLD.

Formula: C₁₇H₁₄O₆
MW: 314.3 g/mol
Melting Pt: 290°C
Storage Temperature: Ambient
MDL Number: MFCD00078140
CAS Number: 7220-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90ad7941dc551259c1647ee169431521f3e8b222&vid=90ad7941dc551259c1647ee169431521f3e8b222&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR